Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012455.2 | NP_036587.2 | 2 | 292 | ||
XM_005263634.2 | XP_005263691.1 | 2 | 357 | |||
XM_006712392.2 | XP_006712455.1 | 2 | 304 | |||
XM_006712393.2 | XP_006712456.1 | 2 | 304 | |||
XM_006712394.2 | XP_006712457.1 | 2 | 304 | |||
GenBank mRNA | AB208798.1 | - | 2 | 295 | ||
AJ459781.1 | - | 1 | 130 | |||
AK290045.1 | - | 2 | 302 | |||
BC035307.1 | - | 2 | 278 | 4339380 | ||
BC073151.1 | - | 2 | 254 | 5757431 | ||
BQ061445.1 | - | 2 | 293 | |||
U63127.1 | - | 1 | 172 |