Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Atg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134394.2 | NP_599221.1 | 4 | 498 | ||
GenBank mRNA | AF175224.1 | - | 4 | 501 | ||
BC078743.1 | - | 4 | 510 | 7112937 |