Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153309.2 | NP_695221.1 | 3, 4 | 597 | ||
XM_008768680.1 | XP_008766902.1 | 3, 4 | 662 | |||
GenBank mRNA | AB083195.1 | - | 2, 3 | 572 | ||
BC087650.1 | - | 3, 4 | 597 | 7310399 |