Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199370.1 | NP_955402.1 | 2, 3 | 527 | ||
GenBank mRNA | AY464139.1 | - | 2, 3 | 527 | ||
BC091106.1 | - | 2, 3 | 585 | 7381703 | ||
BC097497.1 | - | 2, 3 | 550 | 7462062 | ||
M63482.1 | - | 2, 3 | 591 | |||
S76054.1 | - | 2, 3 | 591 |