Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adipor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207587.1 | NP_997470.1 | 8 | 1589 | ||
XM_006249852.2 | XP_006249914.1 | 8 | 1625 | |||
XM_008769543.1 | XP_008767765.1 | 8 | 1926 | |||
XM_008769544.1 | XP_008767766.1 | 8 | 2288 | |||
GenBank mRNA | BC061838.1 | - | 8 | 1589 | 5599938 | |
FQ217287.1 | - | 1 | 371 | |||
FQ233463.1 | - | 1 | 361 |