Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psbpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207593.2 | NP_997476.2 | 2 | 215 | ||
GenBank mRNA | BC099748.1 | - | 2 | 209 | ||
J00776.1 | - | 1 | 127 | |||
V01256.1 | - | 2 | 212 |