Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hyal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207599.2 | NP_997482.2 | 2 | 609 | ||
XM_008766504.1 | XP_008764726.1 | 2 | 608 | |||
GenBank mRNA | AB100602.1 | - | 1 | 473 | ||
BC097259.1 | - | 2 | 609 | 7453545 |