Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323627.1 | NP_001310556.1 | 8, 9 | 1297 | ||
NM_013304.2 | NP_037436.1 | 8, 9 | 1239 | |||
XM_005255915.1 | XP_005255972.1 | 7, 8 | 973 | |||
XM_006721185.1 | XP_006721248.1 | 7, 8 | 973 | |||
XM_011523058.1 | XP_011521360.1 | 7, 8 | 973 | |||
XM_011523059.1 | XP_011521361.1 | 7, 8 | 973 | |||
XM_011523060.1 | XP_011521362.1 | 6, 7 | 772 | |||
GenBank mRNA | BC021908.1 | - | 8, 9 | 1265 | 4053374 | |
BM724650.1 | - | 1, 2 | 103 |