Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258461.1 | NP_001245390.1 | 4, 5 | 869 | ||
NM_006712.4 | NP_006703.1 | 4, 5 | 950 | |||
NM_033015.3 | NP_148936.2 | 3, 4 | 527 | |||
XM_005249932.1 | XP_005249989.1 | 4, 5 | 1142 | |||
XM_005249933.3 | XP_005249990.1 | 4, 5 | 1348 | |||
XM_011515761.2 | XP_011514063.1 | 4, 5 | 950 | |||
XM_011515762.2 | XP_011514064.1 | 4, 5 | 950 | |||
XM_011515763.1 | XP_011514065.1 | 3, 4 | 408 | |||
XM_017011704.1 | XP_016867193.1 | 4, 5 | 1061 | |||
XM_017011705.1 | XP_016867194.1 | 3, 4 | 505 | |||
XM_017011706.1 | XP_016867195.1 | 3, 4 | 697 | |||
GenBank mRNA | AB209783.1 | - | 4, 5 | 770 | ||
AK223132.1 | - | 4, 5 | 860 | |||
AK292232.1 | - | 4, 5 | 835 | |||
AL713798.1 | - | 2, 3 | 1803 | |||
BC000377.2 | - | 4, 5 | 862 | 2820338 | ||
BC006386.2 | - | 3, 4 | 833 | |||
BC011770.2 | - | 4, 5 | 861 | 3831196 | ||
BC039026.2 | - | 3, 4 | 497 | 4940575 | ||
X86779.1 | - | 4, 5 | 854 |