Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000322.4 | NP_000313.2 | 3 | 1527 | ||
GenBank mRNA | M62958.1 | - | 3 | 1489 | ||
M73531.1 | - | 3 | 1489 |