Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324281.1 | NP_001311210.1 | 14, 15 | 2227 | ||
NM_032415.5 | NP_115791.3 | 13, 14 | 2166 | |||
XM_011515586.2 | XP_011513888.1 | 14, 15 | 2195 | |||
XM_011515587.2 | XP_011513889.1 | 13, 14 | 2158 | |||
GenBank mRNA | AF322641.1 | - | 13, 14 | 2068 | ||
AF352576.1 | - | 11, 12 | 1741 | |||
AK074049.1 | - | 13, 14 | 2086 | |||
BC111719.1 | - | 12, 13 | 1852 | 40009168 |