Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039582.3 | NP_001034671.3 | 10 | 1313 | ||
NM_001135740.1 | NP_001129212.1 | 10 | 947 | |||
XM_005274651.2 | XP_005274708.1 | 10 | 930 | |||
XM_006724809.3 | XP_006724872.1 | 10 | 1139 | |||
XM_011531107.2 | XP_011529409.2 | 10 | 1018 | |||
XM_011531108.2 | XP_011529410.1 | 10 | 983 | |||
XM_011531110.2 | XP_011529412.1 | 10 | 1035 | |||
XM_011531111.2 | XP_011529413.1 | 10 | 953 | |||
XM_011531112.2 | XP_011529414.1 | 10 | 926 | |||
XM_017029277.1 | XP_016884766.1 | 10 | 1238 | |||
XM_017029278.1 | XP_016884767.1 | 11 | 1142 | |||
XM_017029279.1 | XP_016884768.1 | 10 | 899 | |||
GenBank mRNA | AK296165.1 | - | 9 | 1356 | ||
AK296203.1 | - | 10 | 1313 | |||
AK303746.1 | - | 10 | 947 | |||
BC064422.1 | - | 9 | 1005 | 5751849 |