Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243466.1 | NP_001230395.1 | 2, 3 | 281 | ||
NM_001243474.1 | NP_001230403.1 | 2, 3 | 311 | |||
NM_001243476.2 | NP_001230405.1 | 6, 7 | 721 | |||
NM_052851.2 | NP_443083.1 | 2, 3 | 311 | |||
NM_178006.3 | NP_821074.1 | 2, 3 | 374 | |||
NM_178007.2 | NP_821075.1 | 2, 3 | 281 | |||
XM_011535299.2 | XP_011533601.1 | 5, 6 | 467 | |||
XM_017020834.1 | XP_016876323.1 | 2, 3 | 479 | |||
XM_017020835.1 | XP_016876324.1 | 4, 5 | 400 | |||
GenBank mRNA | AK091804.1 | - | 2, 3 | 283 | ||
AK094704.1 | - | 2, 4 | 413 | |||
AK308430.1 | - | 2, 3 | 371 | |||
AK308453.1 | - | 2, 3 | 420 | |||
AK312888.1 | - | 2, 3 | 367 | |||
AL049801.1 | - | 2, 3 | 267 | |||
AY082589.1 | - | 2, 3 | 369 | |||
AY082590.1 | - | 2, 3 | 293 | |||
AY082591.1 | - | 2, 3 | 267 | |||
AY082592.1 | - | 2, 3 | 369 | |||
AY366448.1 | - | 2, 3 | 309 | |||
BC046563.1 | - | 2, 3 | 282 | 5527069 | ||
BP233049.1 | - | 2, 3 | 312 | |||
BX647695.1 | - | 6, 7 | 681 | |||
DC402824.1 | - | 2, 3 | 376 |