Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPSAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003294.3 | NP_003285.2 | 6 | 1171 | ||
GenBank mRNA | BC038416.1 | - | 5 | 1125 | ||
BC051852.1 | - | 5 | 1124 | |||
BC065923.1 | - | 5 | 1330 | 5752632 | ||
M30038.1 | - | 5 | 1132 | |||
M33491.1 | - | 5 | 1115 | |||
M37488.1 | - | 5 | 1120 |
Gene Symbol: TPSB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024164.5 | NP_077078.5 | 6 | 1142 | ||
GenBank mRNA | BC028059.1 | - | 5 | 1131 | 5181815 | |
BC029356.1 | - | 6 | 1142 | 4695458 | ||
M33492.1 | - | 5 | 1117 | |||
S55551.1 | - | 5 | 1126 |