Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145134.1 | NP_001138606.1 | 3 | 400 | ||
NM_001145135.1 | NP_001138607.1 | 3 | 400 | |||
NM_001145137.1 | NP_001138609.1 | 2 | 424 | |||
NM_004377.3 | NP_004368.1 | 3 | 361 | |||
NM_152245.2 | NP_689451.1 | 3 | 400 | |||
NM_152246.2 | NP_689452.1 | 3 | 361 | |||
GenBank mRNA | AK223174.1 | - | 3 | 300 | ||
AK291316.1 | - | 3 | 325 | |||
AK297866.1 | - | 3 | 312 | |||
AK299395.1 | - | 3 | 337 | |||
BC142954.1 | - | 2 | 367 | 8860341 | ||
BG426428.1 | - | 346 | ||||
BI560870.1 | - | 2 | 429 | |||
D87812.1 | - | 2 | 281 | |||
DC373481.1 | - | 3 | 400 | |||
U62733.1 | - | 2 | 308 | |||
U66828.1 | - | 3 | 300 | |||
Y08682.1 | - | 3 | 313 | |||
Y08683.1 | - | 3 | 313 |
Gene Symbol: CHKB-CPT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027928.2 | 13 | 1832 | |||
GenBank mRNA | AB051457.1 | - | 10 | 2470 | ||
AK301477.1 | - | 12 | 1399 | |||
BC131570.1 | - | 5 | 657 | 40108355 |