Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005198.4 | NP_005189.2 | 3, 4 | 675 | ||
GenBank mRNA | AB029886.1 | - | 3, 4 | 525 | ||
AK314324.1 | - | 3, 4 | 509 | |||
AL096780.1 | - | 3, 4 | 600 | |||
AL096781.1 | - | 3, 4 | 457 | |||
BC024286.1 | - | 3, 4 | 454 | |||
BC082263.1 | - | 3, 4 | 512 | 6384695 | ||
BC101488.1 | - | 3, 4 | 520 | 8068994 | ||
BC113521.1 | - | 3, 4 | 520 | 8322573 | ||
CR456419.1 | - | 3, 4 | 586 |
Gene Symbol: CHKB-CPT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027928.2 | 3, 4 | 675 | |||
GenBank mRNA | AB051457.1 | - | 1, 2 | 1227 | ||
AK301477.1 | - | 1, 2 | 131 |