Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR2DS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012313.1 | NP_036445.1 | 3 | 265 | ||
GenBank mRNA | AF022047.1 | - | 3 | 258 | ||
AM851002.1 | - | 3 | 290 | |||
AY789063.2 | - | 2 | 238 | |||
L76670.1 | - | 3 | 265 | |||
X97231.1 | - | 3 | 313 |
Gene Symbol: 1060P11.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ630586.1 | - | 4 | 540 |
Gene Symbol: KIR2DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015868.2 | NP_056952.2 | 3 | 291 | ||
GenBank mRNA | AF022048.1 | - | 3 | 254 | ||
AF135566.1 | - | 1 | 191 | |||
AF285435.1 | - | 2 | 214 | |||
AY789057.1 | - | 3 | 254 | |||
BC032422.1 | - | 3 | 267 | 5220339 | ||
BC050730.1 | - | 3 | 285 | 5219336 | ||
BC110613.1 | - | 3 | 268 | 40031619 | ||
FN395076.1 | - | 3 | 254 | |||
FN552550.1 | - | 3 | 236 | |||
L41268.1 | - | 3 | 291 | |||
L76662.1 | - | 3 | 265 | |||
L76663.1 | - | 3 | 265 | |||
U24074.1 | - | 3 | 275 | |||
U50546.1 | - | 3 | 265 | |||
U73395.1 | - | 3 | 275 | |||
U96190.1 | - | 1 | 191 |
Gene Symbol: KIR2DS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291695.1 | NP_001278624.1 | 3 | 289 | ||
NM_001291701.1 | NP_001278630.1 | 3 | 289 | |||
NM_012312.4 | NP_036444.1 | 3 | 289 | |||
GenBank mRNA | AF135565.1 | - | 1 | 191 | ||
AF285438.1 | - | 2 | 214 | |||
AJ002102.1 | - | 3 | 254 | |||
AJ002103.1 | - | 3 | 254 | |||
AJ002104.1 | - | 3 | 254 | |||
AJ002105.1 | - | 3 | 254 | |||
AM409094.1 | - | 3 | 255 | |||
AY366243.1 | - | 3 | 264 | |||
AY366244.1 | - | 3 | 253 | |||
BC108917.1 | - | 3 | 256 | 40017299 | ||
BC119631.1 | - | 3 | 273 | 40115568 | ||
BC119632.1 | - | 3 | 273 | 40115570 | ||
BC125146.1 | - | 2 | 238 | |||
BC126856.1 | - | 3 | 256 | 40017301 | ||
FR750994.1 | - | 3 | 289 | |||
L41347.1 | - | 3 | 273 | |||
L76667.1 | - | 3 | 265 | |||
U24079.1 | - | 3 | 252 | |||
X89893.1 | - | 3 | 254 |
Gene Symbol: KIR3DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242867.1 | NP_001229796.1 | 4 | 572 | ||
NM_006737.3 | NP_006728.2 | 4 | 572 | |||
XM_011526940.2 | XP_011525242.1 | 4 | 696 | |||
XM_017026784.1 | XP_016882273.1 | 4 | 696 | |||
GenBank mRNA | AF208683.1 | - | 2 | 476 | ||
AF208684.1 | - | 2 | 476 | |||
AF208685.1 | - | 2 | 476 | |||
AF208686.1 | - | 2 | 476 | |||
AF208687.1 | - | 2 | 476 | |||
AF208690.1 | - | 2 | 476 | |||
AF262965.1 | - | 2 | 458 | |||
AF262966.1 | - | 2 | 458 | |||
AF262967.1 | - | 4 | 572 | |||
AF263617.1 | - | 4 | 572 | |||
AJ276125.1 | - | 4 | 540 | |||
AW951147.1 | - | 1 | 197 | |||
AY059418.1 | - | 2 | 470 | |||
AY059419.1 | - | 2 | 470 | |||
AY059420.1 | - | 2 | 470 | |||
AY366256.2 | - | 4 | 551 | |||
AY789067.1 | - | 4 | 539 | |||
AY789068.1 | - | 4 | 539 | |||
AY789069.1 | - | 3 | 509 | |||
AY789070.1 | - | 4 | 539 | |||
AY789071.1 | - | 3 | 509 | |||
BC105678.1 | - | 4 | 564 | 40004817 | ||
BC107951.1 | - | 4 | 564 | 40004820 | ||
DQ256504.1 | - | 2 | 471 | |||
EF582384.1 | - | 4 | 539 | |||
L41270.1 | - | 4 | 558 | |||
L76665.1 | - | 4 | 550 | |||
L76666.1 | - | 4 | 550 | |||
U30272.1 | - | 4 | 541 | |||
X93595.1 | - | 4 | 569 | |||
X93596.1 | - | 4 | 539 | |||
X94373.1 | - | 4 | 540 | |||
X94374.1 | - | 4 | 539 |