Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR2DS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF135565.1 | - | 1 | 132 |
Gene Symbol: KIR2DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014219.2 | NP_055034.2 | 3 | 224 | ||
GenBank mRNA | AF285434.1 | - | 2 | 155 | ||
AY366242.1 | - | 3 | 224 | |||
KP260924.1 | - | 3 | 191 | |||
L76669.1 | - | 3 | 206 | |||
U24075.1 | - | 3 | 209 |
Gene Symbol: KIR2DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015868.2 | NP_056952.2 | 3 | 232 | ||
GenBank mRNA | AF022048.1 | - | 3 | 195 | ||
AF135566.1 | - | 1 | 132 | |||
AF135567.1 | - | 1 | 132 | |||
AF285435.1 | - | 2 | 155 | |||
AY789057.1 | - | 3 | 195 | |||
BC032422.1 | - | 3 | 208 | 5220339 | ||
BC050730.1 | - | 3 | 226 | 5219336 | ||
BC110613.1 | - | 3 | 209 | 40031619 | ||
FN395076.1 | - | 3 | 195 | |||
FN552550.1 | - | 3 | 177 | |||
L41268.1 | - | 3 | 232 | |||
L76662.1 | - | 3 | 206 | |||
L76663.1 | - | 3 | 206 | |||
U24074.1 | - | 3 | 216 | |||
U50546.1 | - | 3 | 206 | |||
U73395.1 | - | 3 | 216 | |||
U96190.1 | - | 1 | 132 |