Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR2DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014512.1 | NP_055327.1 | 3 | 267 | ||
GenBank mRNA | AF022046.1 | - | 3 | 267 | ||
AF285437.1 | - | 2 | 214 | |||
AM999888.1 | - | 3 | 262 | |||
X89892.1 | - | 3 | 274 | |||
X98858.1 | - | 3 | 254 |
Gene Symbol: KIR2DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014218.2 | NP_055033.2 | 3 | 292 | ||
XM_011526939.2 | XP_011525241.1 | 3 | 897 | |||
XM_017026782.1 | XP_016882271.1 | 3 | 270 | |||
XM_017026783.1 | XP_016882272.1 | 3 | 897 | |||
GenBank mRNA | AF022045.1 | - | 3 | 261 | ||
AF135560.1 | - | 1 | 191 | |||
AF285431.1 | - | 3 | 287 | |||
AF285432.1 | - | 2 | 214 | |||
AY366237.1 | - | 3 | 290 | |||
AY366238.1 | - | 3 | 236 | |||
AY366239.1 | - | 3 | 284 | |||
AY366240.1 | - | 3 | 292 | |||
AY366241.1 | - | 3 | 291 | |||
AY388471.1 | - | 3 | 284 | |||
AY789055.1 | - | 3 | 287 | |||
AY789056.1 | - | 3 | 254 | |||
BC069344.1 | - | 3 | 287 | 7262296 | ||
KJ699235.1 | - | 3 | 527 | |||
L41267.1 | - | 3 | 286 | |||
U24076.1 | - | 3 | 287 | |||
U24078.1 | - | 3 | 294 | |||
U96189.1 | - | 1 | 191 |