Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002838.4 | NP_002829.3 | 9 | 1028 | ||
NM_080921.3 | NP_563578.2 | 6 | 545 | |||
XM_006711472.3 | XP_006711535.1 | 8 | 845 | |||
XM_006711473.3 | XP_006711536.1 | 8 | 773 | |||
XM_006711474.3 | XP_006711537.1 | 7 | 629 | |||
GenBank mRNA | AK292131.1 | - | 6 | 418 | ||
AK299986.1 | - | 8 | 703 | |||
AK303539.1 | - | 9 | 901 | |||
AK307635.1 | - | 8 | 762 | |||
AK308217.1 | - | 7 | 635 | |||
BC014239.2 | - | 8 | 748 | |||
DA670254.1 | - | 6 | 448 | |||
DA819314.1 | - | 6 | 545 | |||
Y00062.1 | - | 7 | 472 | |||
Y00638.1 | - | 9 | 895 |