Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNLRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281727.1 | NP_001268656.1 | 4 | 363 | ||
NM_001281728.1 | NP_001268657.1 | 5 | 442 | |||
NM_001281729.1 | NP_001268658.1 | 5 | 750 | |||
NM_014183.3 | NP_054902.1 | 4 | 367 | |||
NM_177953.2 | NP_808852.1 | 5 | 446 | |||
NR_104032.1 | 5 | 449 | ||||
GenBank mRNA | AF132750.1 | - | 4 | 347 | ||
AF161511.1 | - | 4 | 338 | |||
AF165516.1 | - | 4 | 320 | |||
AK026864.1 | - | 3 | 303 | |||
BC002481.1 | - | 4 | 345 | 3347555 | ||
BG110161.1 | - | 4 | 352 | |||
BG419592.1 | - | 5 | 418 | |||
BM791453.1 | - | 5 | 428 | |||
BQ639804.1 | - | 5 | 415 | |||
BU163605.1 | - | 386 | ||||
CB149456.1 | - | 4 | 363 |