Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2L11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204107.1 | NP_001191036.1 | 5 | 610 | ||
NM_001204109.1 | NP_001191038.1 | 3 | 704 | |||
NM_001204112.1 | NP_001191041.1 | 4 | 524 | |||
NM_138622.3 | NP_619528.1 | 4 | 808 | |||
NM_138623.3 | NP_619529.1 | 5 | 628 | |||
NM_207003.2 | NP_996886.1 | 4 | 538 | |||
GenBank mRNA | AB071195.1 | - | 3 | 520 | ||
AB071196.1 | - | 4 | 340 | |||
AF455755.1 | - | 2 | 1479 | |||
AY305715.1 | - | 3 | 250 | |||
AY305716.1 | - | 3 | 147 | |||
AY423441.1 | - | 3 | 250 | |||
AY423442.1 | - | 4 | 322 | |||
AY428962.1 | - | 2 | 416 | |||
DQ849202.1 | - | 3 | 236 |