Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165257.1 | NP_001158729.1 | 3 | 551 | ||
NM_013243.3 | NP_037375.2 | 4 | 604 | |||
GenBank mRNA | AF078851.1 | - | 4 | 486 | ||
AF453583.1 | - | 4 | 609 | |||
AK075314.1 | - | 4 | 463 | |||
AK290175.1 | - | 4 | 440 | |||
AK296466.1 | - | 3 | 390 | |||
AY359093.1 | - | 4 | 465 | |||
BC014539.1 | - | 4 | 240 | 4283082 | ||
BI552493.1 | - | 4 | 611 |