Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006983.1 | NP_008914.1 | 7, 8 | 1030 | ||
XM_017002615.1 | XP_016858104.1 | 6, 7 | 1938 | |||
GenBank mRNA | AB010961.1 | - | 7, 8 | 1019 | ||
AB010962.1 | - | 6, 7 | 1111 | |||
AF056200.1 | - | 7, 8 | 1007 | |||
AJ005256.1 | - | 7, 8 | 1030 | |||
AK297700.1 | - | 5, 6 | 1502 | |||
BC025719.1 | - | 7, 8 | 1021 | 5228894 |
Gene Symbol: MMP23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002946.1 | 5, 6 | 699 | |||
GenBank mRNA | AF057061.1 | - | 4, 5 | 673 | ||
AF057062.1 | - | 3, 4 | 625 | |||
AF086353.1 | - | 4, 5 | 445 |