Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256732.1 | NP_001243661.1 | 17 | 1820 | ||
NM_001256733.1 | NP_001243662.1 | 16 | 1736 | |||
NM_001256734.1 | NP_001243663.1 | 16 | 1730 | |||
NM_001256735.1 | NP_001243664.1 | 16 | 1706 | |||
NM_001256736.1 | NP_001243665.1 | 15 | 1607 | |||
NM_012446.3 | NP_036578.2 | 17 | 1796 | |||
XM_011543297.2 | XP_011541599.1 | 16 | 1636 | |||
XM_017009304.1 | XP_016864793.1 | 17 | 1638 | |||
XM_017009305.1 | XP_016864794.1 | 18 | 1727 | |||
XM_017009306.1 | XP_016864795.1 | 16 | 1612 | |||
XM_017009307.1 | XP_016864796.1 | 16 | 1524 | |||
XM_017009308.1 | XP_016864797.1 | 18 | 1899 | |||
XM_017009309.1 | XP_016864798.1 | 17 | 1963 | |||
XM_017009310.1 | XP_016864799.1 | 17 | 2111 | |||
GenBank mRNA | AF161465.1 | - | 17 | 1626 | ||
AK074537.1 | - | 17 | 1591 | |||
AK294864.1 | - | 15 | 1402 | |||
AK297494.1 | - | 17 | 1504 | |||
AL080076.1 | - | 17 | 1644 | |||
BC017020.1 | - | 17 | 1644 | 3913337 | ||
CR936787.1 | - | 16 | 1551 |