Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199756.1 | NP_001186685.1 | 7 | 936 | ||
NM_006243.3 | NP_006234.1 | 7 | 1404 | |||
GenBank mRNA | AK302202.1 | - | 7 | 936 | ||
AK312530.1 | - | 8 | 1219 | |||
BC022474.1 | - | 7 | 990 | 4815091 | ||
BC110883.1 | - | 7 | 1113 | 6045358 | ||
L42373.1 | - | 7 | 1401 |