Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DROSHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100412.1 | NP_001093882.1 | 6 | 1357 | ||
NM_013235.4 | NP_037367.3 | 5 | 1281 | |||
XM_005248291.3 | XP_005248348.1 | 6 | 1357 | |||
XM_005248292.3 | XP_005248349.1 | 6 | 1357 | |||
XM_011514033.2 | XP_011512335.1 | 6 | 1357 | |||
XM_017009400.1 | XP_016864889.1 | 6 | 1357 | |||
GenBank mRNA | AF189011.1 | - | 5 | 1167 | ||
AK308093.1 | - | 5 | 1166 | |||
BC054003.1 | - | 5 | 1169 | 6148714 | ||
BX647724.1 | - | 6 | 1260 |