Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N6AMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013240.5 | NP_037372.4 | 6 | 794 | ||
NM_182749.4 | NP_877426.4 | 5 | 710 | |||
GenBank mRNA | AF139682.1 | - | 6 | 795 | ||
BC011554.1 | - | 5 | 691 | 4640080 |