Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145354.1 | NP_001138826.1 | 14 | 1755 | ||
NM_001321316.1 | NP_001308245.1 | 13 | 1686 | |||
NM_013255.4 | NP_037387.2 | 13 | 1683 | |||
XM_006715993.2 | XP_006716056.1 | 13 | 1683 | |||
GenBank mRNA | AF047489.1 | - | 13 | 1637 | ||
AK002024.1 | - | 13 | 1625 | |||
AK294384.1 | - | 14 | 1756 | |||
AK310452.1 | - | 13 | 1636 | |||
AL162071.1 | - | 4 | 663 | |||
BC002834.2 | - | 8 | 1061 | |||
BC067825.1 | - | 13 | 1640 | 5268939 | ||
BX537433.1 | - | 1 | 35 |