Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013283.4 | NP_037415.1 | 7 | 2001 | ||
NM_182796.2 | NP_877725.1 | 7 | 2140 | |||
GenBank mRNA | AB073390.1 | - | 5 | 3281 | ||
AL136664.1 | - | 7 | 1914 | |||
AY358695.1 | - | 7 | 1894 | |||
DQ395260.1 | - | 4 | 1408 | |||
DQ413182.1 | - | 3 | 1406 | |||
DQ413183.1 | - | 7 | 1928 |