Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013335.3 | NP_037467.2 | 3 | 171 | ||
NM_205847.2 | NP_995319.1 | 3 | 452 | |||
XM_005246483.3 | XP_005246540.1 | 3 | 171 | |||
XM_005246486.3 | XP_005246543.1 | 3 | 140 | |||
XM_011511032.2 | XP_011509334.1 | 3 | 140 | |||
XM_017003906.1 | XP_016859395.1 | 3 | 171 | |||
XM_017003907.1 | XP_016859396.1 | 3 | 140 | |||
XM_017003908.1 | XP_016859397.1 | 3 | 446 | |||
XM_017003909.1 | XP_016859398.1 | 3 | 171 | |||
XM_017003910.1 | XP_016859399.1 | 3 | 140 | |||
XM_017003911.1 | XP_016859400.1 | 3 | 446 | |||
XM_017003912.1 | XP_016859401.1 | 3 | 171 | |||
XM_017003913.1 | XP_016859402.1 | 3 | 140 | |||
XM_017003914.1 | XP_016859403.1 | 3 | 161 | |||
XM_017003915.1 | XP_016859404.1 | 3 | 391 | |||
XM_017003916.1 | XP_016859405.1 | 3 | 161 | |||
XM_017003917.1 | XP_016859406.1 | 3 | 161 | |||
GenBank mRNA | AB209592.1 | - | 2 | 278 | ||
AF135422.2 | - | 2 | 150 | |||
AK000999.1 | - | 3 | 145 | |||
AK022578.1 | - | 3 | 446 | |||
AK222951.1 | - | 3 | 135 | |||
AK290671.1 | - | 3 | 448 | |||
AK296193.1 | - | 3 | 127 | |||
BC007456.2 | - | 3 | 440 | 2989461 | ||
BP284488.1 | - | 3 | 452 | |||
BP366441.1 | - | 3 | 171 |