Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013347.4 | NP_037479.1 | 1 | 666 | ||
GenBank mRNA | BC069791.1 | - | 1 | 561 | 7262610 | |
BC069808.1 | - | 1 | 561 | 7262634 | ||
BC069824.1 | - | 1 | 561 | 7262622 | ||
BC104013.2 | - | 1 | 537 | 40024430 | ||
BC104014.2 | - | 1 | 537 | 40024431 | ||
U24186.2 | - | 1 | 667 |