Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSGIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182981.2 | NP_892026.1 | 5 | 595 | ||
GenBank mRNA | AF191740.1 | - | 3 | 233 | ||
AK290383.1 | - | 5 | 577 | |||
AY037158.1 | - | 5 | 560 | |||
AY258066.1 | - | 6 | 1046 | |||
AY258067.1 | - | 5 | 886 | |||
AY258068.1 | - | 5 | 576 | |||
BC020434.1 | - | 5 | 561 | |||
BC032476.1 | - | 5 | 560 | 5215262 | ||
BC093687.1 | - | 5 | 478 | 7939532 | ||
BC113417.1 | - | 5 | 478 | 8322469 |