Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006658.4 | NP_006649.2 | 3 | 442 | ||
XM_011515094.1 | XP_011513396.1 | 3 | 274 | |||
GenBank mRNA | AF071789.1 | - | 2 | 167 | ||
AF097730.1 | - | 2 | 164 | |||
AK312989.1 | - | 3 | 244 | |||
BC028094.1 | - | 3 | 226 | 5241722 | ||
DA061079.1 | - | 3 | 442 | |||
DB039499.1 | - | 3 | 274 |