Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3IL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271686.1 | NP_001258615.1 | 2, 3 | 490 | ||
NM_013401.3 | NP_037533.2 | 2, 3 | 514 | |||
XM_005274140.4 | XP_005274197.1 | 2, 3 | 479 | |||
XM_006718647.3 | XP_006718710.1 | 2, 3 | 481 | |||
XM_011545196.2 | XP_011543498.1 | 2, 3 | 485 | |||
XM_011545197.2 | XP_011543499.1 | 2, 3 | 801 | |||
XM_011545198.2 | XP_011543500.1 | 2, 3 | 509 | |||
XM_011545199.2 | XP_011543501.1 | 2, 3 | 487 | |||
XM_011545201.2 | XP_011543503.1 | 2, 3 | 492 | |||
GenBank mRNA | AF084557.1 | - | 1, 2 | 68 | ||
BC022239.1 | - | 2, 3 | 430 | 4390573 | ||
BC051820.1 | - | 2, 3 | 490 | 5188805 | ||
BQ946081.1 | - | 2, 3 | 456 | |||
DR004757.1 | - | 2, 3 | 516 |