Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEXI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014015.3 | NP_054734.2 | 2 | 876 | ||
XM_011522455.2 | XP_011520757.1 | 3 | 258 | |||
XM_017023171.1 | XP_016878660.1 | 3 | 1644 | |||
GenBank mRNA | AF108145.2 | - | 2 | 492 | ||
AF129929.1 | - | 2 | 388 | |||
BC001083.2 | - | 2 | 612 | 2958243 | ||
BC037569.1 | - | 2 | 876 | 5227606 | ||
BC052989.1 | - | 2 | 609 | 6194689 | ||
BC071683.1 | - | 2 | 775 | 5190179 |