Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142364.1 | NP_001135836.1 | 4 | 415 | ||
NM_013338.4 | NP_037470.1 | 5 | 505 | |||
XM_017020552.1 | XP_016876041.1 | 4 | 443 | |||
GenBank mRNA | AF102850.1 | - | 5 | 513 | ||
AF161498.1 | - | 5 | 454 | |||
AF183413.1 | - | 5 | 464 | |||
AK299085.1 | - | 4 | 391 | |||
AK299126.1 | - | 5 | 474 | |||
BC012531.1 | - | 5 | 457 | 4273297 | ||
BF668393.1 | - | 479 | ||||
BG482604.1 | - | 5 | 491 | |||
BP246765.1 | - | 5 | 505 | |||
CB988638.1 | - | 502 |