Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013372.6 | NP_037504.1 | 2 | 281 | ||
XM_005254301.2 | XP_005254358.1 | 2 | 345 | |||
XM_017022077.1 | XP_016877566.1 | 2 | 327 | |||
GenBank mRNA | AF045800.1 | - | 1 | 122 | ||
AF110137.2 | - | 2 | 184 | |||
AF154054.1 | - | 2 | 251 | |||
AK312265.1 | - | 2 | 282 | |||
AY232290.1 | - | 2 | 257 | |||
BC069525.1 | - | 1 | 122 | 7262108 | ||
BC093778.1 | - | 2 | 144 | 7939623 | ||
BC101611.1 | - | 2 | 144 | 8069117 |