Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD6IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162429.2 | NP_001155901.1 | 3 | 470 | ||
NM_001256192.1 | NP_001243121.1 | 3 | 470 | |||
NM_013374.5 | NP_037506.2 | 3 | 470 | |||
XM_011533252.1 | XP_011531554.1 | 3 | 693 | |||
XM_011533253.1 | XP_011531555.1 | 3 | 232 | |||
XM_017005488.1 | XP_016860977.1 | 3 | 232 | |||
GenBank mRNA | AF151793.1 | - | 3 | 438 | ||
AF250394.1 | - | 3 | 402 | |||
AF349951.1 | - | 3 | 443 | |||
BC020066.1 | - | 3 | 420 | 4340998 | ||
BC068454.1 | - | 3 | 451 | 4830724 | ||
BT007367.1 | - | 3 | 312 | |||
DA652213.1 | - | 3 | 473 | |||
GQ131806.1 | - | 3 | 396 |