Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMOD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013353.2 | NP_037485.2 | 6 | 792 | ||
XM_011509449.1 | XP_011507751.1 | 6 | 674 | |||
XM_017001089.1 | XP_016856578.1 | 5 | 1409 | |||
XM_017001090.1 | XP_016856579.1 | 6 | 721 | |||
GenBank mRNA | AF165217.1 | - | 6 | 669 | ||
AF177173.2 | - | 6 | 737 | |||
AK092700.1 | - | 6 | 658 | |||
AK300675.1 | - | 4 | 495 | |||
AK313280.1 | - | 6 | 701 | |||
BC017810.1 | - | 6 | 702 | 4288769 | ||
BX647346.1 | - | 5 | 524 |