Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135820.1 | NP_001129292.1 | 18 | 3709 | ||
NM_013390.2 | NP_037522.1 | 19 | 3898 | |||
XM_005251869.4 | XP_005251926.1 | 19 | 3531 | |||
GenBank mRNA | AB037833.2 | - | 19 | 3491 | ||
AF137030.1 | - | 19 | 3506 | |||
AK075370.1 | - | 12 | 1626 | |||
AK303765.1 | - | 2 | 397 | |||
BC140773.1 | - | 18 | 3211 | 9021641 | ||
BC144510.1 | - | 19 | 3366 | |||
BC146780.1 | - | 19 | 3491 | 9007233 | ||
BC171782.1 | - | 18 | 3212 |