Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SACM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319072.1 | NP_001306001.1 | 19 | 2816 | ||
NM_001319073.1 | NP_001306002.1 | 21 | 2984 | |||
NM_014016.4 | NP_054735.3 | 20 | 2944 | |||
XM_011533500.2 | XP_011531802.1 | 20 | 2735 | |||
GenBank mRNA | AB020658.1 | - | 20 | 2788 | ||
AK126434.1 | - | 7 | 2259 | |||
AL136831.1 | - | 20 | 2734 | |||
BC016559.1 | - | 20 | 2740 | 3049075 | ||
BC033581.1 | - | 1 | 1345 | |||
CR749564.1 | - | 20 | 3880 |