Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSX2IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166293.1 | NP_001159765.1 | 14 | 2360 | ||
NM_001166294.1 | NP_001159766.1 | 13 | 2228 | |||
NM_001166295.1 | NP_001159767.1 | 13 | 2356 | |||
NM_001166417.1 | NP_001159889.1 | 15 | 2538 | |||
NM_014021.3 | NP_054740.3 | 15 | 2410 | |||
XM_005270427.1 | XP_005270484.1 | 14 | 2299 | |||
XM_005270428.1 | XP_005270485.1 | 14 | 2257 | |||
XM_005270429.1 | XP_005270486.1 | 14 | 2296 | |||
XM_006710331.1 | XP_006710394.1 | 14 | 2217 | |||
XM_011540610.1 | XP_011538912.1 | 15 | 2307 | |||
XM_017000234.1 | XP_016855723.1 | 14 | 2288 | |||
XM_017000235.1 | XP_016855724.1 | 14 | 2263 | |||
XM_017000236.1 | XP_016855725.1 | 14 | 2279 | |||
XM_017000237.1 | XP_016855726.1 | 13 | 2068 | |||
GenBank mRNA | AB023140.1 | - | 14 | 2348 | ||
AK001710.1 | - | 10 | 1570 | |||
AK292475.1 | - | 14 | 2248 | |||
AK316472.1 | - | 13 | 2134 | |||
AL133046.1 | - | 15 | 2350 | |||
AY367055.1 | - | 15 | 2273 | |||
AY651262.1 | - | 12 | 1929 | |||
BC033637.1 | - | 14 | 2235 | 5578582 | ||
BC035580.1 | - | 13 | 2169 |