Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR10H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013939.2 | NP_039227.1 | 1 | 909 | ||
GenBank mRNA | BC069085.1 | - | 1 | 889 | 7216899 | |
BC069457.1 | - | 1 | 889 | 7262254 | ||
BC112178.1 | - | 1 | 921 | 8327646 | ||
BC113647.1 | - | 1 | 909 | 8322699 |
Set Membership: |
G-protein coupled receptor |