Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014166.3 | NP_054885.1 | 1, 2 | 185 | ||
GenBank mRNA | AF161475.1 | - | 1, 2 | 147 | ||
AF230381.1 | - | 1, 2 | 122 | |||
AF300618.1 | - | 1, 2 | 122 | |||
AK001934.1 | - | 1, 2 | 148 | |||
AK222713.1 | - | 1, 2 | 148 | |||
AK223017.1 | - | 1, 2 | 140 | |||
BC005189.1 | - | 1, 2 | 159 | 3929797 | ||
BP309440.1 | - | 1, 2 | 185 | |||
CR457276.1 | - | 1, 2 | 122 | |||
CR541830.1 | - | 1, 2 | 122 | |||
HG518167.1 | - | 1, 2 | 185 |