Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014041.3 | NP_054760.3 | 4 | 724 | ||
GenBank mRNA | AF092138.1 | - | 4 | 453 | ||
AK027426.1 | - | 4 | 453 | |||
AX775783.1 | - | 4 | 453 | |||
AX775785.1 | - | 5 | 1238 | |||
AX775787.1 | - | 4 | 433 | |||
AX775833.1 | - | 4 | 443 | |||
BC000884.1 | - | 4 | 433 | 3462307 | ||
BI826431.1 | - | 4 | 418 | |||
BQ423345.1 | - | 4 | 460 | |||
L38852.1 | - | 4 | 443 |