Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014080.4 | NP_054799.4 | 10 | 1276 | ||
XM_005254421.2 | XP_005254478.1 | 10 | 1276 | |||
GenBank mRNA | AB209010.1 | - | 7 | 1404 | ||
AF181972.1 | - | 2 | 59 | |||
AF230496.1 | - | 10 | 1258 | |||
AF267981.1 | - | 10 | 1276 |