Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014062.2 | NP_054781.1 | 9 | 1518 | ||
NR_074074.1 | 8 | 1387 | ||||
GenBank mRNA | AB026125.1 | - | 9 | 1476 | ||
AF190161.1 | - | 9 | 1445 | |||
AK001028.1 | - | 9 | 1473 | |||
AK126196.1 | - | 1 | 3003 | |||
AY487344.1 | - | 9 | 1492 | |||
BC000050.2 | - | 6 | 1105 | |||
BC064630.1 | - | 9 | 1461 | 5740890 | ||
BM793847.1 | - | 1 | 202 |