Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014227.2 | NP_055042.1 | 2 | 167 | ||
XM_006724308.3 | XP_006724371.1 | 3 | 418 | |||
XM_011530342.2 | XP_011528644.1 | 4 | 536 | |||
XM_011530343.2 | XP_011528645.1 | 2 | 82 | |||
XM_011530344.2 | XP_011528646.1 | 1 | 27 | |||
XM_017028920.1 | XP_016884409.1 | 4 | 497 | |||
GenBank mRNA | AJ133127.1 | - | 2 | 181 |