Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014314.3 | NP_055129.2 | 1, 2 | 271 | ||
GenBank mRNA | AF038963.1 | - | 1, 2 | 270 | ||
AK301678.1 | - | 1, 2 | 142 | |||
AK315040.1 | - | 1, 2 | 142 | |||
BC107731.1 | - | 1, 2 | 293 | |||
BC132786.1 | - | 1, 2 | 219 | 40146808 | ||
BC136610.1 | - | 1, 2 | 219 | 9020600 | ||
DN998145.1 | - | 1, 2 | 272 |